missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium Bromide 98.0+%, TCI America™

Supplier:  TCI America M0779100G

 View more versions of this product

Catalog No. M0779100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyltriphenylphosphonium Bromide
233°C
C19H18BrP
100 g
LSEFCHWGJNHZNT-UHFFFAOYSA-M
methyltriphenylphosphanium bromide
74505
≥98.0% (HPLC,T)
1779-49-3
White
MFCD00011804
methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
[Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
357.23
357.23
Crystalline Powder

Chemical Identifiers

1779-49-3
357.23
LSEFCHWGJNHZNT-UHFFFAOYSA-M
74505
[Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C19H18BrP
MFCD00011804
methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
methyltriphenylphosphanium bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4293

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions