missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. p-7132356
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M077925G 25 g
M0779100G 100 g
M0779500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M077925G Supplier TCI America Supplier No. M077925G
Only null left

Chemical Identifiers

CAS 1779-49-3
Molecular Formula C19H18BrP
Molecular Weight (g/mol) 357.23
MDL Number MFCD00011804
InChI Key LSEFCHWGJNHZNT-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
PubChem CID 74505
IUPAC Name methyltriphenylphosphanium bromide
SMILES [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 233°C
Color White
Quantity 25 g
Formula Weight 357.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyltriphenylphosphonium Bromide
EINECSNumber (3)-4293
TSCA Yes