missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade

CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol

$72.80 - $183.38

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC394210010
SDS View Documents
Thermo Scientific Chemicals
394210010
1 g Glass bottle
Each for $72.80
Only null left
 
AC394210050
SDS View Documents
Thermo Scientific Chemicals
394210050
5 g Glass bottle
Each for $183.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

1945-77-3
848.779
LGVVZVZPOQJZKT-UHFFFAOYSA-N
2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
C37H44N2Na4O13S
MFCD00148905
131850582
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

1945-77-3
MFCD00148905
LGVVZVZPOQJZKT-UHFFFAOYSA-N
CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]
848.779
844.76
Glass bottle
Red
Crystalline Powder
C37H44N2Na4O13S
Solubility in water: 460g/L (20°C).
Authentic
2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
131850582
ACS Reagent
pH 6.5 (yellow) to pH 8.5 (blue):Pass Test
1 g
Methylthymol blue, tetrasodium salt
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only