missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade
CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol
Supplier: Thermo Scientific Chemicals 394210010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| Methylthymol blue, tetrasodium salt | |
| C37H44N2Na4O13S | |
| Solubility in water: 460g/L (20°C). | |
| Authentic | |
| 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium | |
| 131850582 | |
| ACS Reagent | |
| pH 6.5 (yellow) to pH 8.5 (blue):Pass Test | |
| 1 g |
| 1945-77-3 | |
| MFCD00148905 | |
| LGVVZVZPOQJZKT-UHFFFAOYSA-N | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] | |
| 848.779 | |
| 844.76 | |
| Glass bottle | |
| Red | |
| Crystalline Powder |
Chemical Identifiers
| 1945-77-3 | |
| 848.779 | |
| LGVVZVZPOQJZKT-UHFFFAOYSA-N | |
| 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
| C37H44N2Na4O13S | |
| MFCD00148905 | |
| 131850582 | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
RUO – Research Use Only