missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade

Catalog No. ac394210050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC394210050 5 g Glass bottle
AC394210010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC394210050 Supplier Thermo Scientific Chemicals Supplier No. 394210050
Only null left

CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

Solubility Information Solubility in water: 460g/L (20°C).
Infrared Spectrum Authentic
Formula Weight 844.76
Grade ACS Reagent
Packaging Glass bottle
Suitability pH 6.5 (yellow) to pH 8.5 (blue):Pass Test
Color Red
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Methylthymol blue, tetrasodium salt

RUO – Research Use Only