missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylthiomethyl p-Tolyl Sulfone 98.0+%, TCI America™
Supplier: TCI America M087525G
Specifications
| Methylthiomethyl p-Tolyl Sulfone | |
| 83°C | |
| C9H12O2S2 | |
| 25 g | |
| XAARLLNXZJTFPQ-UHFFFAOYSA-N | |
| 1-methyl-4-(methylsulfanylmethylsulfonyl)benzene | |
| 548421 | |
| ≥98.0% (GC) |
| 59662-65-6 | |
| White | |
| MFCD00010750 | |
| methylthiomethyl p-tolyl sulfone, methyl tosylmethyl sulfane, mt-sulfone, methylmercaptomethyl p-tolyl sulfone, 1-methyl-4-methylsulfanylmethylsulfonyl benzene, 1-methyl-4-methylsulfanyl methanesulfonylbenzene, methylthiomethylp-tolylsulfone, acmc-209mf0, methyl thiomethyl p-tolylsulfone, xaarllnxzjtfpq-uhfffaoysa | |
| CC1=CC=C(C=C1)S(=O)(=O)CSC | |
| 216.313 | |
| 216.31 | |
| Crystalline Powder |
Chemical Identifiers
| 59662-65-6 | |
| 216.313 | |
| XAARLLNXZJTFPQ-UHFFFAOYSA-N | |
| 548421 | |
| CC1=CC=C(C=C1)S(=O)(=O)CSC |
| C9H12O2S2 | |
| MFCD00010750 | |
| methylthiomethyl p-tolyl sulfone, methyl tosylmethyl sulfane, mt-sulfone, methylmercaptomethyl p-tolyl sulfone, 1-methyl-4-methylsulfanylmethylsulfonyl benzene, 1-methyl-4-methylsulfanyl methanesulfonylbenzene, methylthiomethylp-tolylsulfone, acmc-209mf0, methyl thiomethyl p-tolylsulfone, xaarllnxzjtfpq-uhfffaoysa | |
| 1-methyl-4-(methylsulfanylmethylsulfonyl)benzene |
Safety and Handling
TSCA : No