missing translation for 'onlineSavingsMsg'
Learn More

Methylthiomethyl p-Tolyl Sulfone 98.0+%, TCI America™

Catalog No. p-7132418
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08755G 5 g
M087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08755G Supplier TCI America Supplier No. M08755G
Only null left

Chemical Identifiers

CAS 59662-65-6
Molecular Formula C9H12O2S2
Molecular Weight (g/mol) 216.313
MDL Number MFCD00010750
InChI Key XAARLLNXZJTFPQ-UHFFFAOYSA-N
Synonym methylthiomethyl p-tolyl sulfone, methyl tosylmethyl sulfane, mt-sulfone, methylmercaptomethyl p-tolyl sulfone, 1-methyl-4-methylsulfanylmethylsulfonyl benzene, 1-methyl-4-methylsulfanyl methanesulfonylbenzene, methylthiomethylp-tolylsulfone, acmc-209mf0, methyl thiomethyl p-tolylsulfone, xaarllnxzjtfpq-uhfffaoysa
PubChem CID 548421
IUPAC Name 1-methyl-4-(methylsulfanylmethylsulfonyl)benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CSC

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 216.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methylthiomethyl p-Tolyl Sulfone