missing translation for 'onlineSavingsMsg'
Learn More

(Methylthio)acetic acid, 98%

CAS: 2444-37-3 | C3H6O2S | 106.139 g/mol

$69.89 - $718.48

Chemical Identifiers

CAS 2444-37-3
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.139
MDL Number MFCD00075444
InChI Key HGTBAIVLETUVCG-UHFFFAOYSA-N
Synonym methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
PubChem CID 75551
ChEBI CHEBI:47870
IUPAC Name 2-methylsulfanylacetic acid
SMILES CSCC(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2253103
View Documents
Thermo Scientific Chemicals
B2253103
1 g
Each for $69.89
Only null left
 
AAB2253106
View Documents
Thermo Scientific Chemicals
B2253106
5 g
Each for $180.05
Only null left
 
AAB2253114
View Documents
Thermo Scientific Chemicals
B2253114
25 g
Each for $718.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2444-37-3
106.139
HGTBAIVLETUVCG-UHFFFAOYSA-N
75551
2-methylsulfanylacetic acid
C3H6O2S
MFCD00075444
methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
CHEBI:47870
CSCC(=O)O

Specifications

2444-37-3
1.224
>110°C (230°F)
C3H6O2S
MFCD00075444
1740201
HGTBAIVLETUVCG-UHFFFAOYSA-N
2-methylsulfanylacetic acid
75551
106.15
(Methylthio)acetic acid
13°C to 14°C
99°C to 100°C (4 mmHg)
Stench
1.495
1 g
methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio
CSCC(=O)O
106.139
CHEBI:47870
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 219-483-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only