Learn More
(Methylthio)acetic acid, 98%
CAS: 2444-37-3 | C3H6O2S | 106.139 g/mol
Supplier: Thermo Scientific Chemicals B2253106
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (Methylthio)acetic acid | |
| 13°C to 14°C | |
| 99°C to 100°C (4 mmHg) | |
| Stench | |
| 1.495 | |
| 5 g | |
| methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio | |
| CSCC(=O)O | |
| 106.139 | |
| CHEBI:47870 | |
| 98% |
| 2444-37-3 | |
| 1.224 | |
| >110°C (230°F) | |
| C3H6O2S | |
| MFCD00075444 | |
| 1740201 | |
| HGTBAIVLETUVCG-UHFFFAOYSA-N | |
| 2-methylsulfanylacetic acid | |
| 75551 | |
| 106.15 |
Chemical Identifiers
| 2444-37-3 | |
| 106.139 | |
| HGTBAIVLETUVCG-UHFFFAOYSA-N | |
| 75551 | |
| 2-methylsulfanylacetic acid |
| C3H6O2S | |
| MFCD00075444 | |
| methylthio acetic acid, 2-methylthioacetic acid, 2-methylthio acetic acid, acetic acid, methylthio, 2-methylsulfanyl acetic acid, methylsulfenylacetic acid, unii-umv7e1ucux, methylmercaptoacetic acid, umv7e1ucux, acetic acid,2-methylthio | |
| CHEBI:47870 | |
| CSCC(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 219-483-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only