accessibility menu, dialog, popup

UserName

CH3SOHO

CAS RN 2444-37-3

IUPAC Name: 2-(methylsulfanyl)acetic acid
Synonyms: (methylthio)acetic acid methylsulfenylacetic acid
Molecular Weight (g/mol): 106.14
Molecular Formula: C3H6O2S
InChi Key: HGTBAIVLETUVCG-UHFFFAOYSA-N
SMILES: CSCC(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)