missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylsulfoxide, 99.9% min., ACS, MilliporeSigma™
Supplier: Technidata MX14585
Specifications
| Methylsulfoxide | |
| 67-68-5 | |
| Liquid | |
| C2H6OS | |
| MFCD00002089 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.14 |
| For ACS analysis | |
| 4 L | |
| 99.9%, min. | |
| (CH3)2SO | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| ACS/NF/USP |
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Safety and Handling
Recommended Storage : Room Temperature