missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfoxide, 99.9% min., ACS, MilliporeSigma™

Catalog No. p-4382229
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX14586 500 mL
MMX14583 1 L
MMX14585 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MMX14586 Supplier Technidata Supplier No. MX14586
Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Quantity 500 mL
Physical Form Liquid
Assay Percent Range 99.9%, min.
Linear Formula (CH3)2SO
Formula Weight 78.14
Grade ACS/NF/USP
Chemical Name or Material Methylsulfoxide, For ACS analysis
Recommended Storage Room Temperature