missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylparaben, FCC, 99-100.5%, Spectrum™ Chemical
M1187, 99-76-3, C8H8O3
Supplier: Spectrum Chemical Mfg Cor M118725KG
Description
Spectrum™ Chemical Methylparaben, FCC is used as a food preservative and has the E number E218. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 61-68-7 | |
| 0.05% | |
| 125-128°C | |
| C15H15NO2 | |
| HYYBABOKPJLUIN-UHFFFAOYSA-N | |
| 2-[(2,3-dimethylphenyl)amino]benzoic acid | |
| 99 to 101% | |
| Crystalline Powder or Lumps |
| 100% | |
| 0.5% | |
| Poly Pail | |
| 2.5 kg | |
| CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C | |
| 241.29 | |
| BP |
Chemical Identifiers
| 61-68-7 | |
| 241.29 | |
| 2-[(2,3-dimethylphenyl)amino]benzoic acid |
| C15H15NO2 | |
| HYYBABOKPJLUIN-UHFFFAOYSA-N | |
| CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C |