accessibility menu, dialog, popup

UserName

CH3NHHOOCH3

CAS RN 61-68-7

IUPAC Name: 2-[(2,3-dimethylphenyl)amino]benzoic acid
Synonyms: mefenamic acid apomefenamic ac. mefenamico antigen brand of mefenamic acid pms mefenamic acid nu pharm brand of mefenamic acid chemidex brand of mefenamic acid bonabol rowa brand of mefenamic acid mefenaminsaeure
Molecular Weight (g/mol): 241.29
Molecular Formula: C15H15NO2
InChi Key: HYYBABOKPJLUIN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)