missing translation for 'onlineSavingsMsg'
Learn More

Methylparaben, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. 18604527
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604527 25 g Poly Pail
18604528 500 g Poly Pail
18604529 2.5 kg Poly Pail
18604530 12 kg Poly Pail
18604531 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604527 Supplier Spectrum Chemical Mfg Cor Supplier No. M118725GM
Get Up to $500 in Chemicals with Purchase
View all promotions

M1187, 99-76-3, C8H8O3

Spectrum™ Chemical Methylparaben, FCC is used as a food preservative and has the E number E218. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 61-68-7
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
InChI Key HYYBABOKPJLUIN-UHFFFAOYSA-N
IUPAC Name 2-[(2,3-dimethylphenyl)amino]benzoic acid
SMILES CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Loss on Drying 0.5%
Melting Range 125-128°C
Packaging Poly Pail
Quantity 25 g
Percent Purity 99 to 101%
Grade BP
Physical Form Crystalline Powder or Lumps