missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium chloride, 3M (22 wt.%) solution in THF, AcroSeal™

Catalog No. AC252561000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC252561000 100 mL AcroSeal™ Glass bottle
AC252568000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC252561000 Supplier Thermo Scientific Chemicals Supplier No. 252561000
Only null left

CAS: 676-58-4 | CH3ClMg | 74.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 676-58-4 , 109-99-9
Molecular Formula CH3ClMg
Molecular Weight (g/mol) 74.79
MDL Number MFCD00000468
InChI Key RQNMYNYHBQQZSP-UHFFFAOYSA-M
Synonym methylmagnesium chloride, chloromethylmagnesium, chloro methyl magnesium, magnesium, chloromethyl, memgcl, methylmagnesiumchloride, methyl magnesium chloride, unii-m5e1132g4w, ch3mgcl, qmabhcaih@
PubChem CID 12670
IUPAC Name chloro(methyl)magnesium
SMILES C[Mg]Cl

Specifications

Color Brown
Boiling Point 66.0°C
Linear Formula CH3MgCl
Density 1.0100g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 74.79
Percent Purity 22 to 24%
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.01
Quantity 100 mL
Flash Point −17°C
Physical Form Liquid
Chemical Name or Material Methylmagnesium chloride, 22 wt. % Solution in Tetrahydrofuran
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 211-629-7
TSCA TSCA

RUO – Research Use Only