missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium bromide, 1M to 1.5M solution in THF, AcroSeal™

Catalog No. AC377388000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377388000 800 mL AcroSeal™ Glass Bottle
AC377381000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC377388000 Supplier Thermo Scientific Chemicals Supplier No. 377388000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-16-1 , 109-99-9
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent
PubChem CID 6349
IUPAC Name magnesium;carbanide;bromide
SMILES C[Mg]Br

Specifications

Color Brown-Gray to Brown-Purple
Linear Formula CH3MgBr
Concentration 0.95 to 1.20M
Density 1.0000g/mL
Solubility Information Solubility in water: vigorous reaction.
Formula Weight 119.24
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1
Quantity 800 mL
Flash Point −21°C
Physical Form Solution
Chemical Name or Material Methylmagnesium bromide, 1M Solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only