missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylenedisalicylic Acid (mixture of isomers) 90.0+%, TCI America™
$31.68 - $272.16
Chemical Identifiers
| CAS | 27496-82-8 |
|---|---|
| Molecular Formula | C15H12O6 |
| MDL Number | MFCD00016506 |
Chemical Identifiers
| 27496-82-8 | |
| C15H12O6 | |
| MFCD00016506 |
Specifications
| 27496-82-8 | |
| C15H12O6 | |
| 25 g | |
| ≥90.0% (T) | |
| Methylenedisalicylic Acid (mixture of isomers) |
| 245°C | |
| MFCD00016506 | |
| 288.26 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : VO4700000
TSCA : Yes