missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylenedisalicylic Acid (mixture of isomers) 90.0+%, TCI America™
Supplier: TCI America M0221500G
Specifications
| Methylenedisalicylic Acid (mixture of isomers) | |
| 245°C | |
| MFCD00016506 | |
| 288.26 | |
| Crystalline Powder |
| 27496-82-8 | |
| C15H12O6 | |
| 500 g | |
| ≥90.0% (T) |
Chemical Identifiers
| 27496-82-8 | |
| C15H12O6 | |
| MFCD00016506 |
Safety and Handling
RTECSNumber : VO4700000
TSCA : Yes