missing translation for 'onlineSavingsMsg'
Learn More

Methylenedisalicylic Acid (mixture of isomers) 90.0+%, TCI America™

Supplier:  TCI America M022125G

 View more versions of this product

Catalog No. M022125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methylenedisalicylic Acid (mixture of isomers)
245°C
MFCD00016506
288.26
Crystalline Powder
27496-82-8
C15H12O6
25 g
≥90.0% (T)

Chemical Identifiers

27496-82-8
C15H12O6
MFCD00016506
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : VO4700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions