missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ α,β-Methyleneadenosine 5'-triphosphate trisodium salt

Catalog No. 320910
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
320910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 320910 Supplier R&D Systems™ Supplier No. 3209/10*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Non-selective P2 agonist

P2-purinoceptor agonist. Also shown to inhibit adenylate cyclase (Ki = 0.5 mM).

Chemical Identifiers

CAS 1343364-54-4
Molecular Formula C11H15N5Na3O12P3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 571.15
Purity >98%
Molecular Weight (g/mol) 571.155
Chemical Name or Material α, β-Methyleneadenosine 5'-triphosphate trisodium salt
Recommended Storage Store at -20°C