missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride (Stabilized/Certified ACS), Fisher Chemical™

Catalog No. D37500
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D37500 500 mL Glass Bottle
D371 1 L Glass Bottle
D374 4 L Glass Bottle
D3720 20 L Steel Pail
D37200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. D37500 Supplier Thermo Fisher Scientific Supplier No. D37 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Stabilized with preservative. Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Chemical Identifiers

CAS 75-09-2 , 67-56-1 , 110-83-8 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Density 1.33g/cm³
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.5 %
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 350mbar at 20°C
Water 0.02% max.
Boiling Point 39°C
Formula Weight 84.93
Percent Purity ≥99.5%
Grade ACS
Concentration or Composition (by Analyte or Components) Methyl alcohol, Cyclohexene, 2-Methyl-2-butene
Chemical Name or Material Methylene Chloride, Stabilized
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison