missing translation for 'onlineSavingsMsg'
Learn More

Methylene Chloride, NF, 99%, Spectrum™ Chemical

M1266, 75-09-2, CH2Cl2

Supplier:  Spectrum Chemical Mfg Cor M12664LTGLCS4

 View more versions of this product

Catalog No. 18604519


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Methylene Chloride, NF is used as a carrier for pharmaceutical tablet coatings. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Methylene Chloride
100%
4 x 4 L
CH2Cl2
UN1593
ClCCl
84.93
NF
75-09-2
39.8°C
Amber Glass Bottle
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
99%

Chemical Identifiers

75-09-2
84.93
YMWUJEATGCHHMB-UHFFFAOYSA-N
ClCCl
CH2Cl2
MFCD00000881
dichloromethane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions