missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue trihydrate, 95%, pure

CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

Supplier:  Thermo Scientific Chemicals 126751000

 View more versions of this product

Catalog No. AC126751000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylene Blue trihydrate
95%
95%
MFCD00150008
15, 6132
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, insoluble in ether
Authentic
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
104827
373.89
Pure
Green
Crystalline Powder With Metal Luster
100.0°C to 110.0°C
7220-79-3
C16H24ClN3O3S
27, 393
Basic Blue 9 trihydrate, C.I. 52015 trihydrate
XQAXGZLFSSPBMK-UHFFFAOYSA-M
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
373.90
15% max. (105°C)
95%
Glass bottle
100 g

Chemical Identifiers

7220-79-3
373.90
XQAXGZLFSSPBMK-UHFFFAOYSA-M
104827
C16H24ClN3O3S
MFCD00150008
Basic Blue 9 trihydrate, C.I. 52015 trihydrate
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

RTECSNumber : SP5740000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only