missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue trihydrate, 95%, pure

CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

$286.24 - $1,573.02

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9 trihydrate, C.I. 52015 trihydrate
PubChem CID 104827
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC126751000
View Documents Promotion Details
Thermo Scientific Chemicals
126751000
100 g Glass bottle
Each for $286.24
Only null left
 
AC126750010
View Documents Promotion Details
Thermo Scientific Chemicals
126750010
1 kg Glass bottle
Each for $1,573.02
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7220-79-3
373.90
XQAXGZLFSSPBMK-UHFFFAOYSA-M
104827
C16H24ClN3O3S
MFCD00150008
Basic Blue 9 trihydrate, C.I. 52015 trihydrate
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

100.0°C to 110.0°C
95%
MFCD00150008
15, 6132
Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, insoluble in ether
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate
104827
15% max. (105°C)
Pure
Crystalline Powder With Metal Luster
7220-79-3
C16H24ClN3O3S
27, 393
Basic Blue 9 trihydrate, C.I. 52015 trihydrate
XQAXGZLFSSPBMK-UHFFFAOYSA-M
Authentic
373.90
373.89
95%
Green
Methylene Blue trihydrate, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

RTECSNumber : SP5740000

SDS
Documents

Documents

RUO – Research Use Only