missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue Solution I, 0.05 mL (1 drop) = 1 mg/L Sulfide, Ricca Chemical
$475.93
Specifications
| Chemical Name or Material | Methylene Blue Solution I |
|---|---|
| CAS | 7220-79-3 , 7732-18-5 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Color | Blue |