missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagents
Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$102.64 - $625.14
Specifications
| Chemical Name or Material | Methylene Blue Chloride Trihydrate (Basic Blue 9) |
|---|---|
| CAS | 7220-79-3 |
| Molecular Formula | C16H18ClN3S, 3H2O |
| Synonym | 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015 |
| Molecular Weight (g/mol) | 373.8981 |
Specifications
| Methylene Blue Chloride Trihydrate (Basic Blue 9) | |
| C16H18ClN3S, 3H2O | |
| 373.8981 | |
| Glass Bottle | |
| Blue/Green |
| 7220-79-3 | |
| 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015 | |
| Reagent | |
| 1.0001 | |
| Solid |