missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagents
Methylene Blue Chloride Trihydrate (Basic Blue 9), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C611050010C1
Chemical Identifiers
| 7220-79-3 | |
| 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015 | |
| Reagent | |
| 1.0001 | |
| Solid |
| C16H18ClN3S, 3H2O | |
| 373.8981 | |
| Glass Bottle | |
| Blue/Green | |
| Methylene Blue Chloride Trihydrate (Basic Blue 9) |
Specifications
| Methylene Blue Chloride Trihydrate (Basic Blue 9) | |
| C16H18ClN3S, 3H2O | |
| 373.8981 | |
| Glass Bottle | |
| Blue/Green | |
| Solid |
| 7220-79-3 | |
| 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride Trihydrate, Basic Blue 9 Trihydrate, Color Index 52015 | |
| Reagent | |
| 1.0001 | |
| 10 g |