missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, 1% (w/v), Ricca Chemical
$71.67 - $214.90
Specifications
| Chemical Name or Material | Methylene Blue |
|---|---|
| CAS | 7732-18-5,7220-79-3 |
| Percent Purity | 1% W/V |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 1% (w/v) Aqueous Solution |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
4850-4
|
Ricca Chemical Company
48504 |
120 mL | Natural Poly Bottle |
Each for $71.67
|
|
||||
|
4850-16
|
Ricca Chemical Company
485016 |
500 mL | Natural Poly Bottle |
Each for $120.40
|
|
||||
|
4850-32
|
Ricca Chemical Company
485032 |
1 L | Natural Poly Bottle |
Each for $214.90
|
|
||||
Specifications
| Methylene Blue | |
| 1% W/V | |
| 1% (w/v) Aqueous Solution | |
| Blue |
| 7732-18-5,7220-79-3 | |
| Laboratory | |
| Natural Poly Bottle |