missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 1% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 48504

 View more versions of this product

Catalog No. 4850-4


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methylene Blue
1% W/V
1% (w/v) Aqueous Solution
Blue
7732-18-5,7220-79-3
Laboratory
Natural Poly Bottle
120 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions