missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylene Blue, 1% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 485016
Specifications
| Methylene Blue | |
| 1% W/V | |
| 1% (w/v) Aqueous Solution | |
| Blue |
| 7732-18-5,7220-79-3 | |
| Laboratory | |
| Natural Poly Bottle | |
| 500 mL |