missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyldiphenylsilane, 97%
CAS: 776-76-1 | C13H13Si | 197.332 g/mol
$75.55 - $213.60
Chemical Identifiers
| CAS | 776-76-1 |
|---|---|
| Molecular Formula | C13H13Si |
| Molecular Weight (g/mol) | 197.332 |
| MDL Number | MFCD00008255 |
| InChI Key | OKHRRIGNGQFVEE-UHFFFAOYSA-N |
| Synonym | methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-, |
| PubChem CID | 6327660 |
| IUPAC Name | methyl(diphenyl)silicon |
| SMILES | C[Si](C1=CC=CC=C1)C2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0421106
|
Thermo Scientific Chemicals
L0421106 |
5 g |
Each for $75.55
|
|
|||||
|
AAL0421114
|
Thermo Scientific Chemicals
L0421114 |
25 g |
Each for $213.60
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 776-76-1 | |
| 197.332 | |
| OKHRRIGNGQFVEE-UHFFFAOYSA-N | |
| 6327660 | |
| C[Si](C1=CC=CC=C1)C2=CC=CC=C2 |
| C13H13Si | |
| MFCD00008255 | |
| methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-, | |
| methyl(diphenyl)silicon |
Specifications
| 266°C to 267°C | |
| 5 g | |
| MFCD00008255 | |
| 120°C (248°F) | |
| OKHRRIGNGQFVEE-UHFFFAOYSA-N | |
| methyl(diphenyl)silicon | |
| 6327660 | |
| 97% | |
| 1.572 | |
| Methyldiphenylsilane |
| 776-76-1 | |
| C13H13Si | |
| 1867183 | |
| methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-, | |
| C[Si](C1=CC=CC=C1)C2=CC=CC=C2 | |
| 197.332 | |
| 198.34 | |
| Odorless | |
| 1.001 |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-281-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only