missing translation for 'onlineSavingsMsg'
Learn More

Methyldiphenylsilane, 97%

CAS: 776-76-1 | C13H13Si | 197.332 g/mol

Supplier:  Thermo Scientific Chemicals L0421114

 View more versions of this product

Catalog No. AAL0421114


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

266°C to 267°C
776-76-1
C13H13Si
1867183
methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-,
C[Si](C1=CC=CC=C1)C2=CC=CC=C2
197.332
198.34
Odorless
1.001
Methyldiphenylsilane
25 g
MFCD00008255
120°C (248°F)
OKHRRIGNGQFVEE-UHFFFAOYSA-N
methyl(diphenyl)silicon
6327660
97%
1.572

Chemical Identifiers

776-76-1
197.332
OKHRRIGNGQFVEE-UHFFFAOYSA-N
6327660
C[Si](C1=CC=CC=C1)C2=CC=CC=C2
C13H13Si
MFCD00008255
methyldiphenylsilane, diphenylmethylsilane, silane, methyldiphenyl, benzene, 1,1'-methylsilylene bis, methyldiphenylsilyl, methyl diphenyl silicon, methyl-di phenyl silicon, silane, methyldiphenyl-,
methyl(diphenyl)silicon
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-281-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only