accessibility menu, dialog, popup

UserName

H3CSi

CAS RN 776-76-1

IUPAC Name: methyldiphenylsilyl
Synonyms: methyldiphenylsilyl
Molecular Weight (g/mol): 197.33
Molecular Formula: C13H13Si
InChi Key: OKHRRIGNGQFVEE-UHFFFAOYSA-N
SMILES: C[Si](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)