missing translation for 'onlineSavingsMsg'
Learn More

Methylbenzethonium Chloride, 98%, Spectrum™ Chemical

Catalog No. p-7165863
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611760 5 g Poly Bottle
18611762 25 g Poly Bottle
18611759 100 g Poly Bottle
18611761 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611760 Supplier Spectrum Chemical Mfg Cor Supplier No. M12445GM
Only null left

Chemical Identifiers

CAS 25155-18-4

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 5 g
Percent Purity 0.98
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.