missing translation for 'onlineSavingsMsg'
Learn More

Methyl (triphenylphosphoranylidene)acetate, 98%

CAS: 2605-67-6 | C21H19O2P | 334.36 g/mol

$208.52 - $208.52

Chemical Identifiers

CAS 2605-67-6
Molecular Formula C21H19O2P
Molecular Weight (g/mol) 334.36
MDL Number MFCD00008455
InChI Key NTNUDYROPUKXNA-UHFFFAOYSA-N
Synonym methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
PubChem CID 17453
IUPAC Name methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169560250
SDS View Documents
Thermo Scientific Chemicals
169560250
25 g Glass bottle
Each for $208.52
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

2605-67-6
334.36
NTNUDYROPUKXNA-UHFFFAOYSA-N
17453
COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C21H19O2P
MFCD00008455
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate

Specifications

2605-67-6
White
2% max. triphenylphosphine+ triphenylphosphine oxide (HPLC)
C21H19O2P
MFCD00008455
01,112; 01,1081; 08,456; 09,426; 10,363; 13,277
Solubility in water: insoluble
COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
334.36
334.34
Powder
164.0°C to 169.0°C
Authentic
Glass bottle
(C6H5)3P=CHCO2CH3
25 g
methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
NTNUDYROPUKXNA-UHFFFAOYSA-N
methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
17453
98%
Methyl (triphenylphosphoranylidene)acetate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 220-018-4

SDS
Documents

Documents

RUO – Research Use Only