accessibility menu, dialog, popup

UserName

CAS RN 2605-67-6

IUPAC Name: methyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Synonyms: carbomethoxymethylenetriphenylphosphorane
Molecular Weight (g/mol): 334.36
Molecular Formula: C21H19O2P
InChi Key: NTNUDYROPUKXNA-UHFFFAOYSA-N
SMILES: COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)