missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl trans-6-Octadecenoate 97.0+%, TCI America™
$238.72 - $814.34
Chemical Identifiers
| CAS | 14620-36-1 |
|---|---|
| Molecular Formula | C19H36O2 |
| Molecular Weight (g/mol) | 296.495 |
| MDL Number | MFCD00069607 |
| InChI Key | QRTVDKVXAFJVRU-BUHFOSPRSA-N |
| Synonym | trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester |
| PubChem CID | 5366845 |
| IUPAC Name | methyl (E)-octadec-6-enoate |
| SMILES | CCCCCCCCCCCC=CCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2310100MG
|
TCI America
M2310100MG |
100 mg |
Each for $238.72
|
|
|||||
|
M2310500MG
|
TCI America
M2310500MG |
500 mg |
Each for $814.34
|
|
|||||
Chemical Identifiers
| 14620-36-1 | |
| 296.495 | |
| QRTVDKVXAFJVRU-BUHFOSPRSA-N | |
| 5366845 | |
| CCCCCCCCCCCC=CCCCCC(=O)OC |
| C19H36O2 | |
| MFCD00069607 | |
| trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester | |
| methyl (E)-octadec-6-enoate |
Specifications
| 14620-36-1 | |
| Colorless | |
| C19H36O2 | |
| 100 mg | |
| QRTVDKVXAFJVRU-BUHFOSPRSA-N | |
| methyl (E)-octadec-6-enoate | |
| 5366845 | |
| ≥97.0% (GC) | |
| Methyl trans-6-Octadecenoate |
| 19°C | |
| 165°C | |
| MFCD00069607 | |
| trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester | |
| CCCCCCCCCCCC=CCCCCC(=O)OC | |
| 296.495 | |
| 296.50 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : No
Recommended Storage : Freezer