CAS RN 14620-36-1
CAS RN 14620-36-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl trans-6-Octadecenoate 97.0+%, TCI America™
CAS: 14620-36-1 Molecular Formula: C19H36O2 Molecular Weight (g/mol): 296.495 MDL Number: MFCD00069607 InChI Key: QRTVDKVXAFJVRU-BUHFOSPRSA-N Synonym: trans-6-Octadecenoic Acid Methyl Ester, Methyl Petroselaidate, Petroselaidic Acid Methyl Ester PubChem CID: 5366845 IUPAC Name: methyl (E)-octadec-6-enoate SMILES: CCCCCCCCCCCC=CCCCCC(=O)OC