missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thiosalicylate 98.0+%, TCI America™

Catalog No. p-7132889
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18815G 5 g
M188125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18815G Supplier TCI America Supplier No. M18815G
Only null left

Chemical Identifiers

CAS 4892-02-8
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00060678
InChI Key BAQGCWNPCFABAY-UHFFFAOYSA-N
Synonym methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester
PubChem CID 21009
IUPAC Name methyl 2-sulfanylbenzoate
SMILES COC(=O)C1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
Formula Weight 168.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Thiosalicylate
RTECSNumber DH3500000
TSCA No