accessibility menu, dialog, popup

UserName

CAS RN 4892-02-8

IUPAC Name: methyl 2-sulfanylbenzoate
Synonyms: methyl 2-sulfanylbenzoate
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: BAQGCWNPCFABAY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1S
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)