missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Thiocyanate 98.0+%, TCI America™
Supplier: TCI America T0201500ML
Specifications
| Methyl Thiocyanate | |
| Yellow | |
| C2H3NS | |
| 500 mL | |
| methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn | |
| CSC#N | |
| 73.113 | |
| CHEBI:61112 | |
| ≥98.0% (GC) |
| 556-64-9 | |
| 130°C | |
| MFCD00001830 | |
| 1992 | |
| VYHVQEYOFIYNJP-UHFFFAOYSA-N | |
| methyl thiocyanate | |
| 11168 | |
| 73.11 | |
| Liquid |
Chemical Identifiers
| 556-64-9 | |
| 73.113 | |
| VYHVQEYOFIYNJP-UHFFFAOYSA-N | |
| 11168 | |
| methyl thiocyanate |
| C2H3NS | |
| MFCD00001830 | |
| methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn | |
| CHEBI:61112 | |
| CSC#N |
Safety and Handling
RTECSNumber : XL1575000
TSCA : Yes