missing translation for 'onlineSavingsMsg'
Learn More

Methyl Thiocyanate 98.0+%, TCI America™

Catalog No. p-7136141
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T020125ML 25 mL
T0201100ML 100 mL
T0201500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T020125ML Supplier TCI America Supplier No. T020125ML
Only null left

Chemical Identifiers

CAS 556-64-9
Molecular Formula C2H3NS
Molecular Weight (g/mol) 73.113
MDL Number MFCD00001830
InChI Key VYHVQEYOFIYNJP-UHFFFAOYSA-N
Synonym methyl sulfocyanate, methylrhodanid, thiocyanic acid, methyl ester, methyl rhodanate, methylthiokyanat, methane, thiocyanato, thiocyanatomethane, methylthiocyanate, thiocyanic acid methyl ester, ch3scn
PubChem CID 11168
ChEBI CHEBI:61112
IUPAC Name methyl thiocyanate
SMILES CSC#N

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 1992
Formula Weight 73.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Thiocyanate
RTECSNumber XL1575000
TSCA Yes