missing translation for 'onlineSavingsMsg'
Learn More

Methyl Sulfide 99.0+%, TCI America™

Catalog No. p-7132166
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
M043125ML 25 mL
M0431100ML 100 mL
M0431500ML 500 mL
3 options

Catalog No. M043125ML

Supplier: TCI America M043125ML

Only null left

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
IUPAC Name (methylsulfanyl)methane
SMILES CSC

Specifications

Melting Point -98°C
Color Colorless
Boiling Point 37°C
Quantity 25 mL
UN Number 1164
Formula Weight 62.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Sulfide
EINECSNumber (2)-0466
RTECSNumber PV5075000
TSCA Yes