missing translation for 'onlineSavingsMsg'
Learn More

Methyl Sulfide 99.0+%, TCI America™

Catalog No. M0431500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0431500ML 500 mL
M043125ML 25 mL
M0431100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0431500ML Supplier TCI America Supplier No. M0431500ML

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
IUPAC Name (methylsulfanyl)methane
SMILES CSC

Specifications

Melting Point -98°C
Color Colorless
Boiling Point 37°C
Quantity 500 mL
UN Number 1164
Formula Weight 62.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Sulfide
EINECSNumber (2)-0466
RTECSNumber PV5075000
TSCA Yes