missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfide, 99+%, extra pure

Catalog No. p-3737000
Change view
Click to view available options
Quantity:
10 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127760100 10 mL Glass bottle
AC127760010 1 L Glass bottle
AC127760025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC127760100 Supplier Thermo Scientific Chemicals Supplier No. 127760100
Only null left

CAS: 75-18-3 | C2H6S | 62.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
IUPAC Name methylsulfanylmethane
SMILES CSC

Specifications

Melting Point -98.0°C
Color Colorless
Density 0.8460g/mL
Boiling Point 38.0°C
Flash Point −48°C
Infrared Spectrum Authentic
Assay Percent Range 0.2% max. Methylmercaptan (GC)
Packaging Glass bottle
Refractive Index 1.4351
Linear Formula (CH3)2S
Quantity 10 mL
Beilstein 01, 288
Fieser 02,156; 04,190; 05,260; 15,88; 16,84; 17,198
Specific Gravity 0.846
Merck Index 15, 3281
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 62.13
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Methyl sulfide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
EINECSNumber 200-846-2

RUO – Research Use Only