missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Ricinoleate 75.0+%, TCI America™
$70.25 - $220.25
Chemical Identifiers
| CAS | 141-24-2 |
|---|---|
| Molecular Formula | C19H36O3 |
| Molecular Weight (g/mol) | 312.49 |
| MDL Number | MFCD00046712 |
| InChI Key | XKGDWZQXVZSXAO-ADYSOMBNSA-N |
| Synonym | methyl ricinoleate, ricinoleic acid methyl ester, flexricin p-1, methyl ricinolate, unii-90fdr3o96y, r,z-methyl 12-hydroxyoctadec-9-enoate, ricinoleic acid, methyl ester, methyl z,12r-12-hydroxyoctadec-9-enoate, methyl 9z,12r-12-hydroxyoctadec-9-enoate, 9-octadecenoic acid, 12-hydroxy-, methyl ester, 9z,12r |
| PubChem CID | 5354133 |
| IUPAC Name | methyl (9Z,12R)-12-hydroxyoctadec-9-enoate |
| SMILES | CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
R002925ML
|
N/A | 25 mL |
Each for $70.25
|
|
|||||
|
R0029500ML
|
N/A | 500 mL |
Each for $220.25
|
|
|||||
Chemical Identifiers
| C19H36O3 | |
| MFCD00046712 | |
| methyl ricinoleate, ricinoleic acid methyl ester, flexricin p-1, methyl ricinolate, unii-90fdr3o96y, r,z-methyl 12-hydroxyoctadec-9-enoate, ricinoleic acid, methyl ester, methyl z,12r-12-hydroxyoctadec-9-enoate, methyl 9z,12r-12-hydroxyoctadec-9-enoate, 9-octadecenoic acid, 12-hydroxy-, methyl ester, 9z,12r | |
| methyl (9Z,12R)-12-hydroxyoctadec-9-enoate |
Specifications
| 141-24-2 | |
| C19H36O3 | |
| methyl ricinoleate, ricinoleic acid methyl ester, flexricin p-1, methyl ricinolate, unii-90fdr3o96y, r,z-methyl 12-hydroxyoctadec-9-enoate, ricinoleic acid, methyl ester, methyl z,12r-12-hydroxyoctadec-9-enoate, methyl 9z,12r-12-hydroxyoctadec-9-enoate, 9-octadecenoic acid, 12-hydroxy-, methyl ester, 9z,12r | |
| CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)OC | |
| 312.49 | |
| 312.49 | |
| Liquid |
| Yellow | |
| MFCD00046712 | |
| XKGDWZQXVZSXAO-ADYSOMBNSA-N | |
| methyl (9Z,12R)-12-hydroxyoctadec-9-enoate | |
| 5354133 | |
| ≥75.0% (GC) | |
| Methyl Ricinoleate |
Safety and Handling
EINECSNumber : (2)-1332&(2)-1341
TSCA : Yes