accessibility menu, dialog, popup

UserName

H3C(R)HOOOCH3(Z)

CAS RN 141-24-2

IUPAC Name: methyl (9Z,12R)-12-hydroxyoctadec-9-enoate
Synonyms: methyl ricinoleate
Molecular Weight (g/mol): 312.49
Molecular Formula: C19H36O3
InChi Key: XKGDWZQXVZSXAO-ADYSOMBNSA-N
SMILES: CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results