missing translation for 'onlineSavingsMsg'
Learn More

Methyl Ricinoleate 75.0+%, TCI America™

Catalog No. R0029500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0029500ML 500 mL
R002925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0029500ML Supplier TCI America Supplier No. R0029500ML

Chemical Identifiers

CAS 141-24-2
Molecular Formula C19H36O3
Molecular Weight (g/mol) 312.49
MDL Number MFCD00046712
InChI Key XKGDWZQXVZSXAO-ADYSOMBNSA-N
Synonym methyl ricinoleate, ricinoleic acid methyl ester, flexricin p-1, methyl ricinolate, unii-90fdr3o96y, r,z-methyl 12-hydroxyoctadec-9-enoate, ricinoleic acid, methyl ester, methyl z,12r-12-hydroxyoctadec-9-enoate, methyl 9z,12r-12-hydroxyoctadec-9-enoate, 9-octadecenoic acid, 12-hydroxy-, methyl ester, 9z,12r
PubChem CID 5354133
IUPAC Name methyl (9Z,12R)-12-hydroxyoctadec-9-enoate
SMILES CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)OC

Specifications

Color Yellow
Quantity 500 mL
Formula Weight 312.49
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Ricinoleate
EINECSNumber (2)-1332&(2)-1341
TSCA Yes