missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red TS, 0.1% (w/v) Alcoholic Solution, Ricca Chemical
Indicator for Acid-Base Titrations
$149.07 - $303.00
Specifications
| Chemical Name or Material | Methyl Red |
|---|---|
| Name Note | Test Solution |
| CAS | 64-17-5 , 67-56-1 , 63451-28-5 |
| CAS Min % | 98.0, 0.12 |
| CAS Max % | 100.0, 0.13 |
Specifications
| Methyl Red | |
| 64-17-5 , 67-56-1 , 63451-28-5 | |
| 100.0, 0.13 | |
| Indicator | |
| pink (pH 4.2) - yellow (pH 6.2) | |
| Red |
| Test Solution | |
| 98.0, 0.12 | |
| 0.1% (w/v) | |
| Natural Poly Bottle | |
| Alcoholic Solution | |
| Liquid |